Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncf2
Entrez Gene ID
364018
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
-
Chromosome Location
Chr.13: 70226434 - 70259019 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100984.1 NP_001094454.1 8 - 9 686 74
XM_006250013.2 XP_006250075.1 7 - 8 905 74
XM_008769664.1 XP_008767886.1 5 - 6 677 74
GenBank mRNA JN864041.1 - 6 - 7 710 74
JN864042.1 - 6 - 7 710 74
JN864045.1 - 6 - 7 704 74
JN864046.1 - 5 - 6 596 74

View Full Product Details