The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105991.1 | NP_001099461.1 | 1 - 2 | 103 | 69 |
XM_006249924.2 | XP_006249986.1 | 1 - 2 | 977 | 69 |