Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scyl3
Entrez Gene ID
360866
Gene Name
SCY1-like 3 (S. cerevisiae)
Gene Aliases
RGD1308992
Chromosome Location
Chr.13: 82231030 - 82255640 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191828.1 NP_001178757.1 3 - 4 718 69
XM_006250175.2 XP_006250237.1 1 - 2 183 69
XM_006250176.2 XP_006250238.1 2 - 3 208 69
XM_008769660.1 XP_008767882.1 2 - 3 1544 69
XM_008769661.1 XP_008767883.1 2 - 3 1544 69
XM_008769662.1 XP_008767884.1 3 - 4 602 69

View Full Product Details