Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cggbp1
Entrez Gene ID
288353
Gene Name
CGG triplet repeat binding protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 1896149 - 1915603 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105900.1 NP_001099370.1 2 - 3 363 105

View Full Product Details