Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgm5
Entrez Gene ID
679990
Gene Name
phosphoglucomutase 5
Gene Aliases
-
Chromosome Location
Chr.1: 242582591 - 242765837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191953.1 NP_001178882.1 9 - 10 1509 77

View Full Product Details