Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsp90b1
Entrez Gene ID
362862
Gene Name
heat shock protein 90, beta, member 1
Gene Aliases
Grp94, Tra1
Chromosome Location
Chr.7: 27226570 - 27240533 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012197.2 NP_001012197.2 2 - 3 274 121
GenBank mRNA BC081917.1 - 2 - 3 274 121

View Full Product Details