Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc18
Entrez Gene ID
291677
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 18
Gene Aliases
RGD1310237
Chromosome Location
Chr.18: 28466892 - 28501016 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013887.1 NP_001013909.1 7 - 8 910 68
XM_006254546.2 XP_006254608.1 6 - 7 889 68
GenBank mRNA BC079289.1 - 7 - 8 910 68
DQ158861.1 - 6 - 7 843 68
FQ214241.1 - 6 - 7 763 68

View Full Product Details