Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gypc
Entrez Gene ID
364837
Gene Name
glycophorin C (Gerbich blood group)
Gene Aliases
GPC
Chromosome Location
Chr.18: 25312382 - 25344614 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013233.1 NP_001013251.1 1 - 2 254 75
GenBank mRNA AY234182.1 - 1 - 2 254 75
BC103641.1 - 1 - 2 167 75

View Full Product Details