The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053398.1 | NP_445850.1 | 7 - 8 | 1163 | 115 |
GenBank mRNA | BC079378.1 | - | 7 - 8 | 1163 | 115 |