Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpf38a
Entrez Gene ID
298374
Gene Name
pre-mRNA processing factor 38A
Gene Aliases
RGD1311129
Chromosome Location
Chr.5: 128174835 - 128186532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106672.1 NP_001100142.1 1 - 2 329 99

View Full Product Details