Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc48a1
Entrez Gene ID
300191
Gene Name
solute carrier family 48 (heme transporter), member 1
Gene Aliases
HRG-1, Hrg1
Chromosome Location
Chr.7: 139271698 - 139279499 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127456.1 NP_001120928.1 2 - 3 350 58
GenBank mRNA BC105856.1 - 1 - 2 263 58
BC158869.1 - 2 - 3 350 58

View Full Product Details