Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt82
Entrez Gene ID
366991
Gene Name
keratin 82, type II
Gene Aliases
Kb22
Chromosome Location
Chr.7: 143196282 - 143206138 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008803.1 NP_001008803.1 3 - 4 612 69

View Full Product Details