Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ces2g
Entrez Gene ID
291952
Gene Name
carboxylesterase 2G
Gene Aliases
Ces2l, RGD1308358
Chromosome Location
Chr.19: 37025379 - 37034611 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106175.1 NP_001099645.1 5 - 6 862 77
XM_006255448.2 XP_006255510.1 4 - 5 610 77

View Full Product Details