Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pip5k1a
Entrez Gene ID
365865
Gene Name
phosphatidylinositol-4-phosphate 5-kinase, type 1, alpha
Gene Aliases
Pip5k1b
Chromosome Location
Chr.2: 196227669 - 196270949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042621.1 NP_001036086.1 3 - 4 518 74
XM_008761315.1 XP_008759537.1 3 - 4 647 74
XM_008761316.1 XP_008759538.1 3 - 4 644 74
GenBank mRNA AY724476.1 - 3 - 4 518 74

View Full Product Details