Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnma2
Entrez Gene ID
305977
Gene Name
paraneoplastic Ma antigen 2
Gene Aliases
-
Chromosome Location
Chr.15: 43581958 - 43589680 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107272.1 NP_001100742.1 2 - 3 222 121
XM_006252116.2 XP_006252178.1 3 - 4 396 121
XM_008770756.1 XP_008768978.1 2 - 3 247 121

View Full Product Details