Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Heg1
Entrez Gene ID
689710
Gene Name
heart development protein with EGF-like domains 1
Gene Aliases
RGD1564641
Chromosome Location
Chr.11: 70234248 - 70322962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248443.2 XP_006248505.1 11 - 12 3580 86

View Full Product Details