Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spock3
Entrez Gene ID
306404
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Gene Aliases
-
Chromosome Location
Chr.16: 28372067 - 28716354 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107310.1 NP_001100780.1 10 - 11 1276 118
XM_008771117.1 XP_008769339.1 11 - 12 1267 118

View Full Product Details