The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013088.1 | NP_001013106.1 | 6 - 7 | 885 | 96 |
GenBank mRNA | BC079327.1 | - | 6 - 7 | 885 | 96 |