The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039009.1 | NP_001034098.1 | 6 - 7 | 811 | 125 |
XM_008763765.1 | XP_008761987.1 | 6 - 7 | 3713 | 125 | |
XM_008763766.1 | XP_008761988.1 | 7 - 8 | 1009 | 125 | |
XM_008763767.1 | XP_008761989.1 | 7 - 8 | 868 | 125 | |
XM_008763768.1 | XP_008761990.1 | 7 - 8 | 3422 | 125 | |
GenBank mRNA | AY886536.1 | - | 6 - 7 | 811 | 125 |