Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1c19
Entrez Gene ID
307096
Gene Name
aldo-keto reductase family 1, member C19
Gene Aliases
RGD1562954
Chromosome Location
Chr.17: 69653368 - 69675204 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100576.1 NP_001094046.1 2 - 3 236 83
XM_008771866.1 XP_008770088.1 1 - 2 1402 83
XM_008771867.1 XP_008770089.1 2 - 3 1097 83

View Full Product Details