The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100576.1 | NP_001094046.1 | 9 - 10 | 1074 | 114 |
XM_008771866.1 | XP_008770088.1 | 8 - 9 | 2240 | 114 | |
XM_008771867.1 | XP_008770089.1 | 9 - 10 | 1935 | 114 |