Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Morf4l2
Entrez Gene ID
317413
Gene Name
mortality factor 4 like 2
Gene Aliases
LRRG00119
Chromosome Location
Chr.X: 107394468 - 107405564 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257295.1 XP_006257357.1 1 - 2 104 95
XM_006257297.1 XP_006257359.1 1 - 2 104 95
GenBank mRNA FQ212552.1 - 1 - 2 67 95
FQ232538.1 - 1 - 2 67 95

View Full Product Details