Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ociad1
Entrez Gene ID
289590
Gene Name
OCIA domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 37458023 - 37475464 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013874.1 NP_001013896.1 8 - 9 869 68
XM_006250894.2 XP_006250956.1 9 - 10 915 68
GenBank mRNA BC083718.1 - 8 - 9 869 68
FQ213030.1 - 8 - 9 878 68

View Full Product Details