The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003404.1 | NP_001003404.1 | 4 - 5 | 570 | 129 |
XM_006227664.2 | XP_006227726.1 | 5 - 6 | 623 | 129 | |
GenBank mRNA | AJ555485.1 | - | 4 - 5 | 592 | 129 |