Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hif1an
Entrez Gene ID
309434
Gene Name
hypoxia-inducible factor 1, alpha subunit inhibitor
Gene Aliases
-
Chromosome Location
Chr.1: 264309214 - 264319269 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113749.1 NP_001107221.1 7 - 8 1018 107
GenBank mRNA BC158810.1 - 7 - 8 1018 107

View Full Product Details