Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Morn4
Entrez Gene ID
293950
Gene Name
MORN repeat containing 4
Gene Aliases
RGD1307336
Chromosome Location
Chr.1: 261321627 - 261337422 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024975.1 NP_001020146.1 4 - 5 456 106
XM_008760402.1 XP_008758624.1 6 - 7 637 106
GenBank mRNA BC091345.1 - 4 - 5 456 106
EF688599.1 - 4 - 5 456 106

View Full Product Details