Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd7
Entrez Gene ID
363082
Gene Name
programmed cell death 7
Gene Aliases
-
Chromosome Location
Chr.8: 70860671 - 70875471 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108768.1 NP_001102238.1 1 - 2 295 91

View Full Product Details