Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtn4ip1
Entrez Gene ID
309912
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
RGD1563384
Chromosome Location
Chr.20: 48881124 - 48925327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107644.3 NP_001101114.1 6 - 7 1199 107
XM_006256589.2 XP_006256651.1 6 - 7 1220 107
XM_006256590.2 XP_006256652.1 6 - 7 1220 107
XM_008772999.1 XP_008771221.1 7 - 8 888 107
GenBank mRNA BC082054.1 - 7 - 8 1541 107
BC166465.1 - 6 - 7 1203 107

View Full Product Details