Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4g
Entrez Gene ID
689004
Gene Name
C-type lectin domain family 4, member G
Gene Aliases
RGD1562320
Chromosome Location
Chr.12: 2273652 - 2278578 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769016.1 XP_008767238.1 1 - 2 441 86

View Full Product Details