The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108400.2 | NP_001101870.1 | 4 - 5 | 545 | 78 |
GenBank mRNA | BC166884.1 | - | 4 - 5 | 545 | 78 |