Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpped1
Entrez Gene ID
362971
Gene Name
metallophosphoesterase domain containing 1
Gene Aliases
RGD1308244
Chromosome Location
Chr.7: 124682868 - 124751398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130569.1 NP_001124041.1 2 - 3 492 71

View Full Product Details