The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025008.1 | NP_001020179.1 | 3 - 4 | 1700 | 125 |
XM_008765509.1 | XP_008763731.1 | 3 - 4 | 1881 | 125 | |
GenBank mRNA | BC091195.1 | - | 3 - 4 | 1700 | 125 |