Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpaa1
Entrez Gene ID
300046
Gene Name
glycosylphosphatidylinositol anchor attachment 1
Gene Aliases
-
Chromosome Location
Chr.7: 117394367 - 117397950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004240.1 NP_001004240.1 9 - 10 1335 109
GenBank mRNA BC078984.1 - 9 - 10 1335 109

View Full Product Details