The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025066.1 | NP_001020237.1 | 2 - 3 | 268 | 79 |
XM_008765643.1 | XP_008763865.1 | 2 - 3 | 918 | 79 | |
GenBank mRNA | BC083768.1 | - | 2 - 3 | 268 | 79 |