Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srpk1
Entrez Gene ID
361811
Gene Name
SRSF protein kinase 1
Gene Aliases
-
Chromosome Location
Chr.20: 5829412 - 5865886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025726.1 NP_001020897.1 8 - 9 831 145
XM_008772734.1 XP_008770956.1 8 - 9 865 145
GenBank mRNA BC099089.1 - 8 - 9 831 145

View Full Product Details