The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047907.3 | NP_001041372.1 | 3 - 4 | 440 | 79 |
GenBank mRNA | DQ480748.1 | - | 3 - 4 | 444 | 79 |
FM031503.1 | - | 3 - 4 | 440 | 79 | |
FQ229123.1 | - | 3 - 4 | 79 |