Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
363229
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
-
Chromosome Location
Chr.9: 59947565 - 60070549 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750698.3 XP_003750746.1 3 - 4 4502 64
XM_006244936.2 XP_006244998.1 3 - 4 1438 64
XM_006244937.2 XP_006244999.1 4 - 5 1426 64
XM_006244938.2 XP_006245000.1 3 - 4 2456 64
XM_008767152.1 XP_008765374.1 3 - 4 2014 64

View Full Product Details