Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a10
Entrez Gene ID
363229
Gene Name
solute carrier family 39 (zinc transporter), member 10
Gene Aliases
-
Chromosome Location
Chr.9: 59947565 - 60070549 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750698.3 XP_003750746.1 8 - 9 5436 67
XM_006244936.2 XP_006244998.1 8 - 9 2372 67
XM_006244937.2 XP_006244999.1 9 - 10 2360 67
XM_006244938.2 XP_006245000.1 8 - 9 3390 67
XM_008767152.1 XP_008765374.1 8 - 9 2948 67

View Full Product Details