Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox12
Entrez Gene ID
363437
Gene Name
reproductive homeobox 12
Gene Aliases
-
Chromosome Location
Chr.X: 124245478 - 124254623 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024901.1 NP_001020072.1 1 - 2 139 67
XM_008773422.1 XP_008771644.1 1 - 2 241 67
GenBank mRNA DQ058659.1 - 1 - 2 139 67

View Full Product Details