Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox12
Entrez Gene ID
363437
Gene Name
reproductive homeobox 12
Gene Aliases
-
Chromosome Location
Chr.X: 124245478 - 124254623 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024901.1 NP_001020072.1 3 - 4 592 74
GenBank mRNA DQ058659.1 - 3 - 4 592 74

View Full Product Details