Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp5
Entrez Gene ID
308325
Gene Name
NLR family, pyrin domain containing 5
Gene Aliases
Nalp5
Chromosome Location
Chr.1: 71453674 - 71486790 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107474.1 NP_001100944.1 9 - 10 2798 72
XM_008758920.1 XP_008757142.1 7 - 8 2670 72

View Full Product Details