Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-M1-4
Entrez Gene ID
294213
Gene Name
RT1 class I, locus M1, gene 4
Gene Aliases
H2-M11
Chromosome Location
Chr.20: 2388728 - 2390948 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008850.1 NP_001008850.1 2 - 3 343 114

View Full Product Details