Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r54
Entrez Gene ID
691438
Gene Name
vomeronasal 2 receptor, 54
Gene Aliases
-
Chromosome Location
Chr.7: 16931774 - 16966830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241129.1 XP_006241191.1 1 - 2 213 136

View Full Product Details