Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinb9d
Entrez Gene ID
306890
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 9d
Gene Aliases
Serpinb9e
Chromosome Location
Chr.17: 32516679 - 32524336 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107351.1 NP_001100821.1 4 - 5 523 76

View Full Product Details