The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107007.1 | NP_001100477.1 | 7 - 8 | 633 | 62 |
XM_008767820.1 | XP_008766042.1 | 7 - 8 | 833 | 62 | |
GenBank mRNA | BC161867.1 | - | 7 - 8 | 826 | 62 |