Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il17c
Entrez Gene ID
691516
Gene Name
interleukin 17C
Gene Aliases
-
Chromosome Location
Chr.19: 55246926 - 55248678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191115.1 NP_001178044.1 3 - 3 562 84
XM_008772650.1 XP_008770872.1 3 - 3 586 84

View Full Product Details