Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ammecr1
Entrez Gene ID
501539
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
RGD1561004
Chromosome Location
Chr.X: 114129834 - 114233011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002730211.4 XP_002730257.1 6 - 6 3004 117

View Full Product Details