Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a48
Entrez Gene ID
361206
Gene Name
solute carrier family 25, member 48
Gene Aliases
RGD1563703
Chromosome Location
Chr.17: 8568239 - 8608213 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771473.1 XP_008769695.1 8 - 8 1977 153

View Full Product Details