Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt35
Entrez Gene ID
287697
Gene Name
keratin 35, type I
Gene Aliases
Ka30, Krt1-2
Chromosome Location
Chr.10: 88006179 - 88009576 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008820.1 NP_001008820.1 2 - 3 559 165

View Full Product Details