The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009508.1 | NP_001009508.1 | 1 - 1 | 628 | 84 |
GenBank mRNA | AY510284.1 | - | 1 - 1 | 628 | 84 |